Assay Details
| Gene Symbol: Myg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021713.2 | NP_068359.1 | 3 - 4 | 524 | 65 | |
| GenBank mRNA | AF289484.1 | - | 3 - 4 | 516 | 65 | |
| AK002656.1 | - | 3 - 4 | 499 | 65 | ||
| AK004104.1 | - | 3 - 4 | 499 | 65 | ||
| AK013232.1 | - | 3 - 4 | 470 | 65 | ||
| BC023124.1 | - | 2 - 3 | 651 | 65 | ||
| BC028501.1 | - | 3 - 4 | 509 | 5256915 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Myg1
Genomic Map