Assay Details
| Gene Symbol: Myg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021713.2 | NP_068359.1 | 4 - 5 | 673 | 64 | |
| GenBank mRNA | AF289484.1 | - | 4 - 5 | 665 | 64 | |
| AK002656.1 | - | 4 - 5 | 648 | 64 | ||
| AK004104.1 | - | 4 - 5 | 648 | 64 | ||
| AK013232.1 | - | 4 - 5 | 619 | 64 | ||
| BC023124.1 | - | 3 - 4 | 800 | 64 | ||
| BC028501.1 | - | 4 - 5 | 658 | 5256915 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Myg1
Genomic Map