Assay Details
| Gene Symbol: Stub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019719.3 | NP_062693.1 | 2 - 3 | 865 | 58 | |
| GenBank mRNA | AF129086.1 | - | 2 - 3 | 412 | 58 | |
| AK002752.1 | - | 2 - 3 | 422 | 58 | ||
| AK004464.1 | - | 2 - 3 | 413 | 58 | ||
| AK045776.1 | - | 2 - 3 | 414 | 58 | ||
| AK158595.1 | - | 2 - 3 | 449 | 58 | ||
| AK166630.1 | - | 2 - 3 | 431 | 58 | ||
| BC020110.1 | - | 2 - 3 | 407 | 58 | ||
| BC027427.1 | - | 2 - 3 | 865 | 4191866 | 58 | |
| BC038939.1 | - | 2 - 3 | 454 | 2598940 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stub1
Genomic Map