Assay Details
| Gene Symbol: Atxn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016843.3 | NP_058539.2 | 10 - 11 | 1321 | 72 | |
| GenBank mRNA | AK002504.1 | - | 10 - 11 | 1432 | 72 | |
| AK003530.1 | - | 10 - 11 | 1248 | 72 | ||
| AK047170.1 | - | 10 - 11 | 1371 | 72 | ||
| AK077738.1 | - | 10 - 11 | 1324 | 72 | ||
| AK078459.1 | - | 10 - 11 | 1401 | 72 | ||
| AK153271.1 | - | 10 - 11 | 1323 | 72 | ||
| AK161950.1 | - | 1338 | 72 | |||
| AK165941.1 | - | 10 - 11 | 1346 | 72 | ||
| AK166899.1 | - | 10 - 11 | 1406 | 72 | ||
| AK170507.1 | - | 10 - 11 | 1338 | 72 | ||
| AK170751.1 | - | 10 - 11 | 1347 | 72 | ||
| BC002285.1 | - | 4 - 5 | 490 | 72 | ||
| BC016410.1 | - | 10 - 11 | 1331 | 3968016 | 72 | |
| BC046802.1 | - | 10 - 11 | 1457 | 6404607 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atxn10
Genomic Map