Assay Details
| Gene Symbol: Atxn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016843.3 | NP_058539.2 | 1 - 2 | 206 | 106 | |
| GenBank mRNA | AK002504.1 | - | 1 - 2 | 315 | 106 | |
| AK003530.1 | - | 1 - 2 | 133 | 106 | ||
| AK047170.1 | - | 1 - 2 | 256 | 106 | ||
| AK049706.1 | - | 1 - 2 | 250 | 106 | ||
| AK077738.1 | - | 1 - 2 | 208 | 106 | ||
| AK078459.1 | - | 1 - 2 | 286 | 106 | ||
| AK153271.1 | - | 1 - 2 | 208 | 106 | ||
| AK161950.1 | - | 223 | 106 | |||
| AK165941.1 | - | 1 - 2 | 231 | 106 | ||
| AK166899.1 | - | 1 - 2 | 291 | 106 | ||
| AK170507.1 | - | 1 - 2 | 223 | 106 | ||
| AK170751.1 | - | 1 - 2 | 232 | 106 | ||
| BC016410.1 | - | 1 - 2 | 216 | 3968016 | 106 | |
| BC046802.1 | - | 1 - 2 | 342 | 6404607 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atxn10
Genomic Map