Assay Details
| Gene Symbol: Atxn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016843.3 | NP_058539.2 | 3 - 4 | 474 | 85 | |
| GenBank mRNA | AK002504.1 | - | 3 - 4 | 583 | 85 | |
| AK003530.1 | - | 3 - 4 | 401 | 85 | ||
| AK047170.1 | - | 3 - 4 | 524 | 85 | ||
| AK049706.1 | - | 3 - 4 | 518 | 85 | ||
| AK077738.1 | - | 3 - 4 | 476 | 85 | ||
| AK078459.1 | - | 3 - 4 | 554 | 85 | ||
| AK153271.1 | - | 3 - 4 | 476 | 85 | ||
| AK161950.1 | - | 491 | 85 | |||
| AK165941.1 | - | 3 - 4 | 499 | 85 | ||
| AK166899.1 | - | 3 - 4 | 559 | 85 | ||
| AK170507.1 | - | 3 - 4 | 491 | 85 | ||
| AK170751.1 | - | 3 - 4 | 500 | 85 | ||
| BC016410.1 | - | 3 - 4 | 484 | 3968016 | 85 | |
| BC046802.1 | - | 3 - 4 | 610 | 6404607 | 85 | |
| X61506.1 | - | 3 - 4 | 588 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atxn10
Genomic Map