Assay Details
| Gene Symbol: Atxn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016843.3 | NP_058539.2 | 8 - 9 | 1083 | 72 | |
| GenBank mRNA | AK003530.1 | - | 8 - 9 | 1010 | 72 | |
| AK047170.1 | - | 8 - 9 | 1133 | 72 | ||
| AK049706.1 | - | 8 - 9 | 1127 | 72 | ||
| AK077738.1 | - | 8 - 9 | 1086 | 72 | ||
| AK078459.1 | - | 8 - 9 | 1163 | 72 | ||
| AK153271.1 | - | 8 - 9 | 1085 | 72 | ||
| AK161950.1 | - | 1100 | 72 | |||
| AK165941.1 | - | 8 - 9 | 1108 | 72 | ||
| AK166899.1 | - | 8 - 9 | 1168 | 72 | ||
| AK170507.1 | - | 8 - 9 | 1100 | 72 | ||
| AK170751.1 | - | 8 - 9 | 1109 | 72 | ||
| AK195052.1 | - | 3 - 4 | 318 | 72 | ||
| BC002285.1 | - | 2 - 3 | 252 | 72 | ||
| BC016410.1 | - | 8 - 9 | 1093 | 3968016 | 72 | |
| BC046802.1 | - | 8 - 9 | 1219 | 6404607 | 72 | |
| X61506.1 | - | 8 - 9 | 1197 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atxn10
Genomic Map