Assay Details
| Gene Symbol: Cdc14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122989.1 | NP_001116461.1 | 10 - 11 | 1056 | 71 | |
| NM_172587.3 | NP_766175.3 | 10 - 11 | 1320 | 71 | ||
| GenBank mRNA | AK041155.1 | - | 10 - 11 | 1042 | 71 | |
| AK044332.1 | - | 10 - 11 | 1622 | 71 | ||
| AK161345.1 | - | 10 - 11 | 1321 | 71 | ||
| BC049794.1 | - | 10 - 11 | 1262 | 6514098 | 71 | |
| BC057357.1 | - | 10 - 11 | 1211 | 6835123 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdc14b
Genomic Map