Assay Details
Gene Symbol: Cdc14b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK161345.1 | - | 12 - 13 | 1585 | 56 | |
AK183075.1 | - | 1 - 2 | 38 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
