| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300014190.mm.1 |
TC1300002198.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300014185.mm.1 |
NM_001122989.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300014185.mm.1 |
NM_001122989.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300014185.mm.1 |
NM_172587.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300014185.mm.1 |
NM_172587.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300014185.mm.1 |
TC1300002198.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300014186.mm.1 |
NM_001122989.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300014186.mm.1 |
NM_001122989.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300014186.mm.1 |
NM_172587.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300014186.mm.1 |
NM_172587.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300014186.mm.1 |
TC1300002198.mm.1 |
MTA TC |