| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200005012.mm.1 |
NM_027597.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005012.mm.1 |
NM_027597.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005012.mm.1 |
NM_029069.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005012.mm.1 |
NM_029069.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005012.mm.1 |
TC1200000710.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200005014.mm.1 |
NM_027597.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005014.mm.1 |
NM_027597.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005014.mm.1 |
NM_029069.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005014.mm.1 |
NM_029069.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005014.mm.1 |
TC1200000710.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200005016.mm.1 |
NM_027597.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005016.mm.1 |
NM_027597.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005016.mm.1 |
NM_029069.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005016.mm.1 |
NM_029069.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005016.mm.1 |
TC1200000710.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200005017.mm.1 |
NM_027597.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005017.mm.1 |
NM_027597.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005017.mm.1 |
NM_029069.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005017.mm.1 |
NM_029069.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005017.mm.1 |
TC1200000710.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200005013.mm.1 |
NM_027597.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005013.mm.1 |
NM_027597.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005013.mm.1 |
NM_029069.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005013.mm.1 |
NM_029069.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005013.mm.1 |
TC1200000710.mm.1 |
MTA TC |