Assay Details
| Gene Symbol: Morf4l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168225.1 | NP_001161697.1 | 1 - 2 | 212 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001168225.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR0X00019011.mm.1 | NM_001168225.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019011.mm.1 | NM_001168225.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019011.mm.1 | NM_001168226.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019011.mm.1 | NM_001168226.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019011.mm.1 | NM_001168227.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019011.mm.1 | NM_001168227.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019011.mm.1 | NM_001168228.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019011.mm.1 | NM_001168228.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019011.mm.1 | NM_001168229.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019011.mm.1 | NM_001168229.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019011.mm.1 | NM_001168230.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019011.mm.1 | NM_001168230.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019011.mm.1 | NM_019768.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019011.mm.1 | NM_019768.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019011.mm.1 | TC0X00003038.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00019013.mm.1 | NM_001168225.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019013.mm.1 | NM_001168225.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019013.mm.1 | NM_001168226.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019013.mm.1 | NM_001168226.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019013.mm.1 | NM_001168227.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019013.mm.1 | NM_001168227.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019013.mm.1 | NM_001168228.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019013.mm.1 | NM_001168228.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019013.mm.1 | NM_001168229.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019013.mm.1 | NM_001168229.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019013.mm.1 | NM_001168230.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019013.mm.1 | NM_001168230.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019013.mm.1 | NM_019768.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019013.mm.1 | NM_019768.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019013.mm.1 | TC0X00003038.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00019018.mm.1 | NM_001168225.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019018.mm.1 | NM_001168225.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019018.mm.1 | NM_001168226.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019018.mm.1 | NM_001168226.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019018.mm.1 | NM_001168227.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019018.mm.1 | NM_001168227.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019018.mm.1 | NM_001168228.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019018.mm.1 | NM_001168228.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019018.mm.1 | NM_001168229.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019018.mm.1 | NM_001168229.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019018.mm.1 | NM_019768.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019018.mm.1 | NM_019768.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019018.mm.1 | TC0X00003038.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00019015.mm.1 | NM_001168225.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019015.mm.1 | NM_001168225.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019015.mm.1 | NM_001168226.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019015.mm.1 | NM_001168226.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019015.mm.1 | NM_001168227.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019015.mm.1 | NM_001168227.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019015.mm.1 | NM_001168228.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019015.mm.1 | NM_001168228.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019015.mm.1 | NM_001168230.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019015.mm.1 | NM_001168230.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019015.mm.1 | NM_019768.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019015.mm.1 | NM_019768.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019015.mm.1 | TC0X00003038.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00019021.mm.1 | NM_001168225.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019021.mm.1 | NM_001168225.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019021.mm.1 | TC0X00003038.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00019022.mm.1 | NM_001168225.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019022.mm.1 | NM_001168225.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019022.mm.1 | TC0X00003038.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00019014.mm.1 | NM_001168225.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019014.mm.1 | NM_001168225.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019014.mm.1 | NM_001168226.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019014.mm.1 | NM_001168226.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019014.mm.1 | NM_001168227.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019014.mm.1 | NM_001168227.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019014.mm.1 | NM_001168228.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019014.mm.1 | NM_001168228.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019014.mm.1 | NM_001168229.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019014.mm.1 | NM_001168229.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019014.mm.1 | NM_001168230.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019014.mm.1 | NM_001168230.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019014.mm.1 | NM_019768.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019014.mm.1 | NM_019768.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019014.mm.1 | TC0X00003038.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00019016.mm.1 | NM_001168225.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019016.mm.1 | NM_001168225.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019016.mm.1 | NM_001168226.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019016.mm.1 | NM_001168226.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019016.mm.1 | NM_001168227.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019016.mm.1 | NM_001168227.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019016.mm.1 | NM_001168228.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019016.mm.1 | NM_001168228.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019016.mm.1 | NM_001168230.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019016.mm.1 | NM_001168230.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019016.mm.1 | NM_019768.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019016.mm.1 | NM_019768.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00019016.mm.1 | TC0X00003038.mm.1 | MTA TC |

Morf4l2
Genomic Map