Assay Details
| Gene Symbol: Rps6kl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146244.4 | NP_666356.2 | 8 - 9 | 1466 | 60 | |
| GenBank mRNA | AK036734.1 | - | 2 - 3 | 1004 | 60 | |
| AK044050.1 | - | 8 - 9 | 1457 | 60 | ||
| BC027298.1 | - | 8 - 9 | 1420 | 3672127 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rps6kl1
Genomic Map