| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200005789.mm.1 |
NM_001252582.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005789.mm.1 |
NM_001252582.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005789.mm.1 |
NM_001252583.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005789.mm.1 |
NM_001252583.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005789.mm.1 |
NM_144833.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005789.mm.1 |
NM_144833.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005789.mm.1 |
NR_045546.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005789.mm.1 |
NR_045546.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005789.mm.1 |
NR_045615.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005789.mm.1 |
NR_045615.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005789.mm.1 |
TC1200000808.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200005791.mm.1 |
NM_001252582.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005791.mm.1 |
NM_001252582.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005791.mm.1 |
NM_001252583.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005791.mm.1 |
NM_001252583.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005791.mm.1 |
NM_144833.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005791.mm.1 |
NM_144833.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005791.mm.1 |
NR_045546.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005791.mm.1 |
NR_045546.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005791.mm.1 |
NR_045615.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005791.mm.1 |
NR_045615.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005791.mm.1 |
TC1200000808.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200005796.mm.1 |
NM_001252582.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005796.mm.1 |
NM_001252582.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005796.mm.1 |
NM_001252583.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005796.mm.1 |
NM_001252583.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005796.mm.1 |
NM_144833.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005796.mm.1 |
NM_144833.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005796.mm.1 |
NR_045615.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005796.mm.1 |
NR_045615.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005796.mm.1 |
TC1200000808.mm.1 |
MTA TC |