Assay Details
| Gene Symbol: Zfp410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144833.3 | NP_659082.1 | 9 - 10 | 1593 | 64 | |
| NR_045546.1 | 9 - 10 | 1561 | 64 | |||
| NR_045615.1 | 7 - 8 | 1277 | 64 | |||
| GenBank mRNA | AF295806.1 | - | 9 - 10 | 1355 | 64 | |
| AK051939.1 | - | 7 - 8 | 993 | 64 | ||
| AK083003.1 | - | 9 - 10 | 1336 | 64 | ||
| AK132345.1 | - | 9 - 10 | 1331 | 64 | ||
| BC021528.1 | - | 9 - 10 | 1348 | 5353673 | 64 | |
| BC056957.1 | - | 9 - 10 | 1290 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp410
Genomic Map