Assay Details
| Gene Symbol: Isca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC022589.1 | - | 1 - 2 | 109 | 4037176 | 74 |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Isca2| Gene Symbol: Isca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC022589.1 | - | 1 - 2 | 109 | 4037176 | 74 |
Set Membership: |
|