Assay Details
| Gene Symbol: Isca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028863.1 | NP_083139.1 | 2 - 3 | 206 | 58 | |
| GenBank mRNA | AK002936.1 | - | 2 - 3 | 205 | 58 | |
| AK019988.1 | - | 1 - 2 | 373 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Isca2
Genomic Map