| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10397269 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10397272 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10397268 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10397271 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10397273 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10397270 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10397274 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1424154_PM_a_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10397267 |
- |
- |
| Mouse Genome 430 2.0 Array |
1424154_a_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1424154_PM_a_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
74316_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
74316_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1200005923.mm.1 |
NM_028863.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005923.mm.1 |
NM_028863.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005923.mm.1 |
TC1200000819.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1424154_PM_a_at |
- |
- |
| Mouse Expression Set 430 |
1424154_a_at |
- |
- |
| Murine Genome U74v2 Set |
96021_at |
- |
- |
| Murine Genome U74v2 Set |
96021_PM_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1424154_a_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1424154_PM_a_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1424154_a_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1424154_PM_a_at |
- |
- |