Assay Details
| Gene Symbol: Cdkn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007669.4 | NP_031695.1 | 1 - 2 | 94 | 62 | |
| GenBank mRNA | AK007630.1 | - | 1 - 2 | 92 | 62 | |
| AK089093.1 | - | 1 - 2 | 92 | 62 | ||
| AK139306.1 | - | 1 - 2 | 95 | 62 | ||
| AK149910.1 | - | 1 - 2 | 95 | 62 | ||
| AK170467.1 | - | 1 - 2 | 95 | 62 | ||
| BC002043.1 | - | 1 - 2 | 47 | 3495942 | 62 | |
| U24173.1 | - | 1 - 2 | 85 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007630.1, AK089093.1, AK139306.1, AK149910.1, AK170467.1, BC002043.1, NM_007669.4, U24173.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Citations (1)
Meis1 is an essential and rate-limiting regulator of MLL leukemia stem cell potential.
- Authors
- Wong P, Iwasaki M, Somervaille TC, So CW, Cleary ML
- Journal
- Genes Dev - Page No 2762 (Thu Nov 01 00:00:00 UTC 2007)
- Volume (Year)
- 21 (2007)
- Product Usage
- -

Cdkn1a
Genomic Map