Assay Details
| Gene Symbol: Rab5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK089199.1 | - | 1 - 2 | 112 | 77 | |
| AK134691.1 | - | 1 - 2 | 51 | 77 | ||
| AK159832.1 | - | 1 - 2 | 101 | 77 | ||
| AK167599.1 | - | 1 - 2 | 101 | 77 | ||
| AK170540.1 | - | 1 - 2 | 121 | 77 | ||
| AK172328.1 | - | 1 - 2 | 58 | 77 | ||
| BC023027.1 | - | 1 - 2 | 67 | 5369566 | 77 | |
| BC029678.1 | - | 1 - 2 | 51 | 5368061 | 77 | |
| BY081527.1 | - | 1 - 2 | 136 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK089199.1, AK159832.1, AK167599.1, AK170540.1, AK172328.1, BC023027.1, BC029678.1 transcripts.
Set Membership: |
|

Rab5c
Genomic Map