Assay Details
Gene Symbol: Rab5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB232595.1 | - | 2 - 3 | 319 | 76 | |
AK089199.1 | - | 3 - 4 | 521 | 76 | ||
AK159832.1 | - | 3 - 4 | 510 | 76 | ||
AK167599.1 | - | 3 - 4 | 510 | 76 | ||
AK170540.1 | - | 3 - 4 | 530 | 76 | ||
AK172328.1 | - | 3 - 4 | 467 | 76 | ||
BC023027.1 | - | 3 - 4 | 476 | 5369566 | 76 | |
BC027378.1 | - | 2 - 3 | 282 | 76 | ||
BC029678.1 | - | 3 - 4 | 460 | 5368061 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
