Assay Details
| Gene Symbol: Rab5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB232595.1 | - | 3 - 4 | 440 | 66 | |
| AK134691.1 | - | 4 - 5 | 581 | 66 | ||
| AK159832.1 | - | 4 - 5 | 631 | 66 | ||
| AK167599.1 | - | 4 - 5 | 631 | 66 | ||
| AK170540.1 | - | 4 - 5 | 651 | 66 | ||
| AK172328.1 | - | 4 - 5 | 588 | 66 | ||
| BC023027.1 | - | 4 - 5 | 597 | 5369566 | 66 | |
| BC027378.1 | - | 3 - 4 | 403 | 66 | ||
| BC029678.1 | - | 4 - 5 | 581 | 5368061 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab5c
Genomic Map