| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100011730.mm.1 |
NM_001035854.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011730.mm.1 |
NM_001035854.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011730.mm.1 |
NM_027915.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011730.mm.1 |
NM_027915.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011730.mm.1 |
TC1100001247.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100011732.mm.1 |
NM_001035854.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011732.mm.1 |
NM_001035854.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011732.mm.1 |
NM_027915.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011732.mm.1 |
NM_027915.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011732.mm.1 |
TC1100001247.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100011731.mm.1 |
NM_001035854.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011731.mm.1 |
NM_001035854.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011731.mm.1 |
NM_027915.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011731.mm.1 |
NM_027915.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011731.mm.1 |
TC1100001247.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100011733.mm.1 |
NM_001035854.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011733.mm.1 |
NM_001035854.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011733.mm.1 |
NM_027915.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011733.mm.1 |
NM_027915.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100011733.mm.1 |
TC1100001247.mm.1 |
MTA TC |