Assay Details
| Gene Symbol: Tefm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183275.2 | NP_899098.2 | 1 - 2 | 74 | 59 | |
| GenBank mRNA | AK146639.1 | - | 1 - 2 | 75 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tefm
Genomic Map