Assay Details
| Gene Symbol: Tefm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183275.2 | NP_899098.2 | 2 - 3 | 528 | 91 | |
| GenBank mRNA | AK146639.1 | - | 2 - 3 | 529 | 91 | |
| BC132029.1 | - | 1 - 2 | 168 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tefm
Genomic Map