Assay Details
| Gene Symbol: Tefm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183275.2 | NP_899098.2 | 3 - 4 | 681 | 140 | |
| GenBank mRNA | AK003325.1 | - | 3 - 4 | 658 | 140 | |
| AK146639.1 | - | 3 - 4 | 682 | 140 | ||
| BC094625.1 | - | 1 - 2 | 115 | 1515023 | 140 | |
| BC132029.1 | - | 2 - 3 | 321 | 140 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK003325.1, BC094625.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P374292 | NM_183275.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P374292 | AK146639.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P374292 | NM_183275.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P374292 | AK146639.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 726029 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 739871 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 511040 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 78013 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132853 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P374292 | NM_183275.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P374292 | AK146639.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P374292 | NM_183275.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P374292 | AK146639.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952805 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952814 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952793 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952797 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952816 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952802 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952809 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952826 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952795 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952791 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952812 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952830 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952823 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27952835 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07412714 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132853 | - | - |
| Whole Mouse Genome G4122A | A_52_P374292 | NM_183275.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P374292 | AK146639.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P374292 | NM_183275.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P374292 | AK146639.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P432468 | AK003325.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134337 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133535 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132853 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09345123 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09345121 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09345119 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09345117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27952826 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27952780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27952783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09080482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27952807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09185865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27952794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27952806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27952800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27952785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27952819 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27952813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27952793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08998688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27952776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27952802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09345120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27952773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27952795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27952814 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09191438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27952772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09648617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27952805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27952812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27952777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27952791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09632573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27952788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09699251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08996254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27952769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09114239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27952809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27952808 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10389009 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389008 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389006 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389007 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452256_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10389005 | - | - |
| Mouse Genome 430 2.0 Array | 1452256_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452256_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68550_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68550_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100030724.mm.1 | NM_183275.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100030724.mm.1 | NM_183275.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100030724.mm.1 | TC1100003340.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100030725.mm.1 | NM_183275.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100030725.mm.1 | NM_183275.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100030725.mm.1 | TC1100003340.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452256_at | - | - |
| Mouse Expression Set 430 | 1452256_PM_at | - | - |
| Murine Genome U74v2 Set | 169511_PM_at | - | - |
| Murine Genome U74v2 Set | 169511_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452256_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452256_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452256_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452256_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 673538 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2850736 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2850736 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002850736 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1216357 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1216357 | - | - |
| mouse-6 v1.1 (Illumina) | scl39855.4.1_72-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2850736 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39855.4.1_72-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2850736 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2938179 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2938179 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl39855.4.1_72-S | - | - |

Tefm
Genomic Map