Assay Details
| Gene Symbol: Mief1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178719.5 | NP_848834.2 | 2 - 3 | 447 | 53 | |
| GenBank mRNA | AK038466.1 | - | 2 - 3 | 420 | 53 | |
| AK041184.1 | - | 2 - 3 | 426 | 53 | ||
| AK050546.1 | - | 2 - 3 | 408 | 53 | ||
| AK080375.1 | - | 2 - 3 | 393 | 53 | ||
| AK163389.1 | - | 3 - 4 | 548 | 53 | ||
| BC094597.1 | - | 2 - 3 | 379 | 53 | ||
| BC137633.1 | - | 1 - 2 | 351 | 8860652 | 53 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK038466.1, AK050546.1, AK080375.1, BC137633.1, NM_178719.5 transcripts.
Set Membership: |
|

Mief1
Genomic Map