Assay Details
| Gene Symbol: Cyp2d34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145474.2 | NP_663449.2 | 4 - 5 | 721 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp2d34| Gene Symbol: Cyp2d34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145474.2 | NP_663449.2 | 4 - 5 | 721 | 70 | |
Set Membership: |
|