Assay Details
| Gene Symbol: Lemd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146075.2 | NP_666187.2 | 1 - 2 | 774 | 81 | |
| GenBank mRNA | AK082131.1 | - | 1 - 2 | 581 | 81 | |
| AK143224.1 | - | 1 - 2 | 759 | 81 | ||
| AY640307.1 | - | 1 - 2 | 754 | 81 | ||
| BC026588.1 | - | 1 - 2 | 84 | 4972900 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK082131.1, BC026588.1 transcripts.
Set Membership: |
|

Lemd2
Genomic Map