Assay Details
| Gene Symbol: Pdzrn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164593.1 | NP_001158065.1 | 7 - 8 | 1410 | 69 | |
| NM_001164594.1 | NP_001158066.1 | 5 - 6 | 847 | 69 | ||
| GenBank mRNA | AK135524.1 | - | 5 - 6 | 830 | 69 | |
| BC151034.1 | - | 7 - 8 | 1339 | 9087947 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdzrn4
Genomic Map