Assay Details
| Gene Symbol: Gsdma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021347.4 | NP_067322.1 | 8 - 9 | 846 | 89 | |
| GenBank mRNA | AB033595.1 | - | 8 - 9 | 835 | 89 | |
| AK028645.1 | - | 8 - 9 | 847 | 89 | ||
| AK028698.1 | - | 8 - 9 | 847 | 89 | ||
| AK029062.1 | - | 8 - 9 | 830 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P314107 | AK029062.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P314107 | AK028645.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P314107 | AK028698.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P314107 | NM_021347.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P314107 | AB033595.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P314107 | AK029062.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P314107 | AK028645.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P314107 | AK028698.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P314107 | NM_021347.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P314107 | AB033595.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 912272 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 684014 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 369499 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 552422 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 360368 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 425501 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 34280 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 26066 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129263 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140205 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P314107 | AK029062.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P314107 | AK028645.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P314107 | AK028698.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P314107 | NM_021347.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P314107 | AB033595.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P314107 | AK029062.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P314107 | AK028645.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P314107 | AK028698.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P314107 | NM_021347.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P314107 | AB033595.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28057695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28057714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28057721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28057706 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28057708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28057685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28057724 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28057713 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28057707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28057698 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28057799 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140205 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02406808 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129263 | - | - |
| Whole Mouse Genome G4122A | A_52_P94941 | AK029062.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P94941 | AK080067.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P94941 | AK028645.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P94941 | AK028698.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P94941 | NM_021347.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P314107 | AK029062.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P314107 | AK028645.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P314107 | AK028698.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P314107 | NM_021347.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P314107 | AB033595.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P314107 | AK029062.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P314107 | AK028645.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P314107 | AK028698.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P314107 | NM_021347.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P314107 | AB033595.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130337 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140205 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129263 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09585415 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09625571 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09585412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28057820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09386758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09716567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09585411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09146317 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057697 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09736025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057712 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09137187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057718 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057699 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057698 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09168179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09678448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057695 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08986265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09392735 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09708963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09486307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09678443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09678450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28057706 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10380985 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380978 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380983 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380977 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380982 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380981 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380979 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380984 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380980 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423634_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1457777_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10380976 | - | - |
| Mouse Genome 430 2.0 Array | 1423634_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423634_at | - | - |
| Mouse Genome 430 2.0 Array | 1457777_at | - | - |
| Mouse Genome 430 2.0 Array | 1457777_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57911_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57911_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100014638.mm.1 | NM_021347.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014638.mm.1 | NM_021347.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014638.mm.1 | TC1100001577.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100014641.mm.1 | NM_021347.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014641.mm.1 | NM_021347.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014641.mm.1 | TC1100001577.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100014642.mm.1 | NM_021347.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014642.mm.1 | NM_021347.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014642.mm.1 | TC1100001577.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100014639.mm.1 | NM_021347.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014639.mm.1 | NM_021347.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014639.mm.1 | TC1100001577.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100014640.mm.1 | NM_021347.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014640.mm.1 | NM_021347.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014640.mm.1 | TC1100001577.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100014643.mm.1 | NM_021347.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014643.mm.1 | NM_021347.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014643.mm.1 | TC1100001577.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100014637.mm.1 | NM_021347.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014637.mm.1 | NM_021347.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014637.mm.1 | TC1100001577.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423634_PM_at | - | - |
| Mouse Expression Set 430 | 1423634_at | - | - |
| Mouse Expression Set 430 | 1457777_PM_at | - | - |
| Mouse Expression Set 430 | 1457777_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423634_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423634_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423634_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423634_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457777_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457777_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 699008 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4230450 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4230450 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004230450 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1213351 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1213351 | - | - |
| mouse-6 v1.1 (Illumina) | scl057911.12_253-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4230450 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl057911.12_253-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4230450 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1213351 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1213351 | - | - |

Gsdma
Genomic Map