Assay Details
| Gene Symbol: Ttc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028918.2 | NP_083194.2 | 1 - 2 | 281 | 69 | |
| GenBank mRNA | AK016657.1 | - | 1 - 2 | 212 | 69 | |
| AK161541.1 | - | 1 - 2 | 220 | 69 | ||
| AK162855.1 | - | 1 - 2 | 221 | 69 | ||
| BC050805.1 | - | 1 - 2 | 305 | 6771545 | 69 | |
| CA463990.1 | - | 1 - 2 | 308 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttc25
Genomic Map