Assay Details
| Gene Symbol: Ttc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028918.2 | NP_083194.2 | 6 - 7 | 1017 | 121 | |
| GenBank mRNA | AK161541.1 | - | 6 - 7 | 956 | 121 | |
| AK162855.1 | - | 6 - 7 | 957 | 121 | ||
| BC050805.1 | - | 6 - 7 | 1041 | 6771545 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ttc25
Genomic Map