Assay Details
| Gene Symbol: Ttc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028918.2 | NP_083194.2 | 9 - 10 | 1505 | 97 | |
| GenBank mRNA | AK016657.1 | - | 9 - 10 | 1436 | 97 | |
| AK136690.1 | - | 3 - 4 | 350 | 97 | ||
| AK161541.1 | - | 9 - 10 | 1444 | 97 | ||
| AK162855.1 | - | 9 - 10 | 1445 | 97 | ||
| BC050805.1 | - | 9 - 10 | 1527 | 6771545 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ttc25
Genomic Map