Assay Details
| Gene Symbol: Rnf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172448.3 | NP_766036.2 | 5 - 6 | 1196 | 71 | |
| GenBank mRNA | AK028750.1 | - | 4 - 5 | 555 | 71 | |
| AK032782.1 | - | 4 - 5 | 1239 | 71 | ||
| AK153638.1 | - | 4 - 5 | 2474 | 71 | ||
| BC075707.1 | - | 3 - 4 | 332 | 71 | ||
| BC131957.1 | - | 3 - 4 | 314 | 40130234 | 71 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rnf43
Genomic Map