Assay Details
| Gene Symbol: Rnf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172448.3 | NP_766036.2 | 7 - 8 | 1467 | 86 | |
| GenBank mRNA | AK028750.1 | - | 6 - 7 | 826 | 86 | |
| AK032782.1 | - | 6 - 7 | 1510 | 86 | ||
| BC075707.1 | - | 5 - 6 | 603 | 86 | ||
| BC131957.1 | - | 5 - 6 | 585 | 40130234 | 86 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rnf43
Genomic Map