Assay Details
| Gene Symbol: Dnal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028821.3 | NP_083097.2 | 4 - 5 | 272 | 111 | |
| GenBank mRNA | AK005826.1 | - | 4 - 5 | 358 | 111 | |
| BC070454.1 | - | 229 | 111 | |||
| BC125392.1 | - | 3 - 4 | 254 | 40129907 | 111 | |
| BC125394.1 | - | 3 - 4 | 250 | 40129909 | 111 | |
More Information
Set Membership: |
|

Dnal1
Genomic Map