Assay Details
| Gene Symbol: Dnal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028821.3 | NP_083097.2 | 6 - 7 | 464 | 141 | |
| GenBank mRNA | AK005826.1 | - | 6 - 7 | 550 | 141 | |
| BC070454.1 | - | 421 | 141 | |||
| BC125392.1 | - | 5 - 6 | 446 | 40129907 | 141 | |
| BC125394.1 | - | 5 - 6 | 442 | 40129909 | 141 | |
| BY126068.1 | - | 3 - 4 | 261 | 142 | ||
More Information
Set Membership: |
|

Dnal1
Genomic Map