Assay Details
| Gene Symbol: Ltbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013589.3 | NP_038617.3 | 8 - 9 | 2006 | 144 | |
| GenBank mRNA | AK052980.1 | - | 8 - 9 | 2003 | 144 | |
| BC119785.1 | - | 8 - 9 | 1990 | 40099725 | 144 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ltbp2
Genomic Map