Assay Details
| Gene Symbol: Arg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009705.3 | NP_033835.1 | 4 - 5 | 609 | 79 | |
| GenBank mRNA | AF032466.1 | - | 4 - 5 | 561 | 79 | |
| BC023349.1 | - | 4 - 5 | 573 | 5254031 | 79 | |
| DV039756.1 | - | 4 - 5 | 643 | 79 | ||
| U90886.1 | - | 4 - 5 | 581 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arg2
Genomic Map