| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700015536.mm.1 |
NM_139154.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700015536.mm.1 |
NM_139154.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700015536.mm.1 |
TC1700001679.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700015535.mm.1 |
NM_139154.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700015535.mm.1 |
NM_139154.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700015535.mm.1 |
TC1700001679.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700015532.mm.1 |
NM_139154.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700015532.mm.1 |
NM_139154.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700015532.mm.1 |
TC1700001679.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700015537.mm.1 |
NM_139154.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700015537.mm.1 |
NM_139154.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700015537.mm.1 |
TC1700001679.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700015544.mm.1 |
NM_139154.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700015544.mm.1 |
NM_139154.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700015544.mm.1 |
TC1700001679.mm.1 |
MTA TC |