Assay Details
| Gene Symbol: Ankrd33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK044543.1 | - | 1 - 2 | 103 | 63 | |
| FJ895380.1 | - | 1 - 2 | 75 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK044543.1, FJ895380.1 transcripts.
Set Membership: |
|

Ankrd33
Genomic Map