Assay Details
| Gene Symbol: Fam109b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177391.4 | NP_796365.1 | 1 - 2 | 95 | 133 | |
| GenBank mRNA | AK083325.1 | - | 1 - 2 | 68 | 134 | |
| BY775160.1 | - | 1 - 2 | 75 | 133 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK083325.1, NM_177391.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382879 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382877 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382878 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P128293 | AK083325.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P128293 | NM_177391.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972648 | BC116256.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972648 | BC116257.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972648 | AK090060.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972648 | AK083325.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972648 | NM_177391.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 42268 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 313477 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 514283 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108105 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972648 | BC116256.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972648 | BC116257.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972648 | AK090060.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972648 | AK083325.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972648 | NM_177391.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P128293 | AK083325.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P128293 | NM_177391.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382892 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382885 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03083959 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108105 | - | - |
| Whole Mouse Genome G4122A | A_52_P305683 | BC116256.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P305683 | BC116257.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P305683 | AK090060.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P305683 | AK083325.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P305683 | NM_177391.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P137875 | BC116257.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P137875 | AK090060.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P137875 | AK083325.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P137875 | NM_177391.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128293 | AK083325.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128293 | NM_177391.4 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108105 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12459613 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12239625 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12004191 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12239624 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382877 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382879 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12033254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12004191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382904 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382881 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382902 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382907 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12239624 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382895 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382899 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12384534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12004201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30382874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12004196 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10425752 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425751 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425754 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10425753 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438202_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10425749 | - | - |
| Mouse Genome 430 2.0 Array | 1438202_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438202_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 338368_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 338368_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500005920.mm.1 | NM_177391.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500005920.mm.1 | NM_177391.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500005920.mm.1 | TC1500002335.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500005916.mm.1 | NM_177391.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500005916.mm.1 | NM_177391.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500005916.mm.1 | TC1500002335.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500005918.mm.1 | NM_177391.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500005918.mm.1 | NM_177391.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500005918.mm.1 | TC1500002335.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500005921.mm.1 | NM_177391.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500005921.mm.1 | NM_177391.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500005921.mm.1 | TC1500002335.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500005915.mm.1 | NM_177391.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500005915.mm.1 | NM_177391.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500005915.mm.1 | TC1500002335.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1438202_at | - | - |
| Mouse Expression Set 430 | 1438202_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438202_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438202_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438202_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438202_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 594215 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 7040411 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 7040411 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007040411 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1234704 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1234704 | - | - |
| mouse-6 v1.1 (Illumina) | scl0338368.2_77-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7040411 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0338368.2_77-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7040411 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2877835 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2877835 | - | - |

Fam109b
Genomic Map