Assay Details
| Gene Symbol: Phf21b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081166.2 | NP_001074635.2 | 6 - 7 | 1163 | 100 | |
| NR_030731.1 | 6 - 7 | 1163 | 100 | |||
| GenBank mRNA | AK042863.1 | - | 6 - 7 | 754 | 100 | |
| AK046530.1 | - | 6 - 7 | 1088 | 100 | ||
| BC067021.1 | - | 6 - 7 | 807 | 100 | ||
| BC157925.1 | - | 6 - 7 | 785 | 9088098 | 100 | |
| BC158044.1 | - | 6 - 7 | 785 | 9007348 | 100 | |
| BC172019.1 | - | 6 - 7 | 785 | 9054305 | 100 | |
| BC172145.1 | - | 6 - 7 | 785 | 9054815 | 100 | |
More Information
Set Membership: |
|

Phf21b
Genomic Map