Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500015815.mm.1 |
NM_001081166.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015815.mm.1 |
NM_001081166.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015815.mm.1 |
NR_030731.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015815.mm.1 |
NR_030731.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015815.mm.1 |
TC1500001993.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500015816.mm.1 |
NM_001081166.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015816.mm.1 |
NM_001081166.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015816.mm.1 |
NR_030731.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015816.mm.1 |
NR_030731.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015816.mm.1 |
TC1500001993.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500015814.mm.1 |
NM_001081166.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015814.mm.1 |
NM_001081166.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015814.mm.1 |
NR_030731.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015814.mm.1 |
NR_030731.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015814.mm.1 |
TC1500001993.mm.1 |
MTA TC |