Assay Details
| Gene Symbol: Phf21b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081166.2 | NP_001074635.2 | 8 - 9 | 1290 | 102 | |
| GenBank mRNA | AK042863.1 | - | 8 - 9 | 881 | 102 | |
| BC067021.1 | - | 8 - 9 | 934 | 102 | ||
| BC157925.1 | - | 8 - 9 | 912 | 9088098 | 102 | |
| BC158044.1 | - | 8 - 9 | 912 | 9007348 | 102 | |
| BC172019.1 | - | 8 - 9 | 912 | 9054305 | 102 | |
More Information
Set Membership: |
|

Phf21b
Genomic Map