Assay Details
| Gene Symbol: Ccdc177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008423.2 | NP_001008423.2 | 1 - 2 | 318 | 86 | |
| GenBank mRNA | AK147473.1 | - | 145 | 86 | ||
| BC067044.1 | - | 1 - 2 | 290 | 30531780 | 86 | |
| BY727768.1 | - | 1 - 2 | 323 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK147473.1, BC067044.1, NM_001008423.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577481 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577471 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577466 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577488 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577465 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577489 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577478 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577474 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577480 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577487 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577468 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577467 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577475 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577476 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577482 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577473 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577472 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577490 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577469 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577470 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990200 | NM_001008423.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990200 | AK147473.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1990200 | BC067044.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 754271 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 45970 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170014 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990200 | NM_001008423.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990200 | AK147473.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1990200 | BC067044.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577486 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577473 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577488 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577474 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577512 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577477 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577492 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28577491 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170014 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170014 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10093148 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10093147 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10093140 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10093129 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09941735 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577489 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577480 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577488 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577465 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577481 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577474 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577478 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577487 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577475 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577468 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577472 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577470 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577473 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577490 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577466 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577482 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577467 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577476 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577469 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28577471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28577521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577491 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577520 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577498 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577526 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577492 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10263161 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577444 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577524 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577508 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577395 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577399 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09865830 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10093140 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10093130 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577449 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577459 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577494 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577500 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577502 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577435 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10049707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577480 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10357680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10077332 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09941735 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10260552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577464 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10168902 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09890309 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577451 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10434144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10093139 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577390 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10182326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577462 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577517 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577472 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577434 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577465 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10162850 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577448 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577488 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577487 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577496 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577398 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577454 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577483 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10357675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10263166 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09890306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28577504 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10401288 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10401287 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460625_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440181_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10401286 | - | - |
| Mouse Genome 430 2.0 Array | 1460625_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440181_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1460625_at | - | - |
| Mouse Genome 430 2.0 Array | 1440181_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 380768_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 380768_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200013801.mm.1 | NM_001008423.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200013801.mm.1 | NM_001008423.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200013801.mm.1 | TC1200002006.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200013805.mm.1 | NM_001008423.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200013805.mm.1 | NM_001008423.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200013805.mm.1 | TC1200002006.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200013803.mm.1 | NM_001008423.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200013803.mm.1 | NM_001008423.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200013803.mm.1 | TC1200002006.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200013806.mm.1 | NM_001008423.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200013806.mm.1 | NM_001008423.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200013806.mm.1 | TC1200002006.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200013807.mm.1 | NM_001008423.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200013807.mm.1 | NM_001008423.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200013807.mm.1 | TC1200002006.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200013804.mm.1 | NM_001008423.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200013804.mm.1 | NM_001008423.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200013804.mm.1 | TC1200002006.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1460625_at | - | - |
| Mouse Expression Set 430 | 1440181_PM_at | - | - |
| Mouse Expression Set 430 | 1440181_at | - | - |
| Mouse Expression Set 430 | 1460625_PM_at | - | - |
| Murine Genome U74v2 Set | 166550_PM_i_at | - | - |
| Murine Genome U74v2 Set | 166550_i_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440181_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1460625_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1460625_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440181_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 609596 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1253471 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2863621 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2863621 | - | - |

Ccdc177
Genomic Map