| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300020500.mm.1 |
NM_198658.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020500.mm.1 |
NM_198658.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020500.mm.1 |
NM_201619.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020500.mm.1 |
NM_201619.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020500.mm.1 |
NM_201622.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020500.mm.1 |
NM_201622.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020500.mm.1 |
TC0300002620.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300020502.mm.1 |
NM_198658.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020502.mm.1 |
NM_198658.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020502.mm.1 |
NM_201619.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020502.mm.1 |
NM_201619.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020502.mm.1 |
NM_201622.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020502.mm.1 |
NM_201622.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020502.mm.1 |
TC0300002620.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300020499.mm.1 |
NM_198658.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020499.mm.1 |
NM_198658.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020499.mm.1 |
NM_201619.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020499.mm.1 |
NM_201619.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020499.mm.1 |
TC0300002620.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300020501.mm.1 |
NM_198658.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020501.mm.1 |
NM_198658.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020501.mm.1 |
NM_201619.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020501.mm.1 |
NM_201619.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020501.mm.1 |
NM_201622.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020501.mm.1 |
NM_201622.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020501.mm.1 |
TC0300002620.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300020498.mm.1 |
NM_198658.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020498.mm.1 |
NM_198658.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020498.mm.1 |
NM_201619.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020498.mm.1 |
NM_201619.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020498.mm.1 |
NM_201622.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020498.mm.1 |
NM_201622.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020498.mm.1 |
TC0300002620.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300020503.mm.1 |
NM_198658.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020503.mm.1 |
NM_198658.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020503.mm.1 |
NM_201619.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020503.mm.1 |
NM_201619.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020503.mm.1 |
NM_201622.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020503.mm.1 |
NM_201622.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300020503.mm.1 |
TC0300002620.mm.1 |
MTA TC |