Assay Details
| Gene Symbol: Pdzrn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164593.1 | NP_001158065.1 | 4 - 5 | 1153 | 65 | |
| NM_001164594.1 | NP_001158066.1 | 2 - 3 | 590 | 65 | ||
| GenBank mRNA | AK135524.1 | - | 2 - 3 | 573 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdzrn4
Genomic Map