Assay Details
| Gene Symbol: Efcab6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161628.1 | NP_001155100.1 | 5 - 6 | 767 | 78 | |
| NM_001161629.1 | NP_001155101.1 | 3 - 4 | 255 | 78 | ||
| NM_029946.4 | NP_084222.4 | 22 - 23 | 2874 | 78 | ||
| GenBank mRNA | AK016516.1 | - | 3 - 4 | 360 | 78 | |
| AK019850.1 | - | 5 - 6 | 767 | 78 | ||
| AK220412.1 | - | 5 - 6 | 686 | 78 | ||
| BC056955.1 | - | 10 - 11 | 1277 | 6812632 | 78 | |
| BC065111.1 | - | 22 - 23 | 2874 | 30536704 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Efcab6
Genomic Map